2-Benzyl-2,9-diazaspiro[5.5]undecane

Lieferant: Apollo Scientific

OR470848-1G
APOSOR470848-1GEA 1540 EUR
APOSOR470848-1G
2-Benzyl-2,9-diazaspiro[5.5]undecane
2-Benzyl-2,9-diazaspiro[5.5]undecane
Formel: C₁₆H₂₄N₂
Molecular Weight: 244,375 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11223505
CAS-Nummer: 867006-13-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR