2-Benzyl-2,6-diazaspiro[4.5]decane
Lieferant: Apollo Scientific
OR470849-1G
APOSOR470849-1GEA
1420
EUR
APOSOR470849-1G
2-Benzyl-2,6-diazaspiro[4.5]decane
2-Benzyl-2,6-diazaspiro[4.5]decane
Formel:
C₁₅H₂₂N₂ Molecular Weight: 230,349 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11223525 CAS-Nummer: 1086395-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...