2-Cbz-2,7-diazaspiro[4.5]decane

Lieferant: Apollo Scientific

OR470850-1G
APOSOR470850-1GEA 2020 EUR
APOSOR470850-1G
2-Cbz-2,7-diazaspiro[4.5]decane
2-Cbz-2,7-diazaspiro[4.5]decane
Formel: C₁₆H₂₂N₂O₂
Molecular Weight: 274,358 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11223504
CAS-Nummer: 1086394-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR