1-(4-Bromobenzyl)-3-[(tert-butyldimethylsilyl)oxy]azetidine
Lieferant: Apollo Scientific
OR470853-1G
APOSOR470853-1GEA
713
EUR
APOSOR470853-1G
1-(4-Bromobenzyl)-3-[(tert-butyldimethylsilyl)oxy]azetidine
1-(4-Bromobenzyl)-3-[(tert-butyldimethylsilyl)oxy]azetidine
Formel:
C₁₆H₂₆BrNOSi Lagertemperatur: Raumtemperatur |
MDL:
MFCD27935369 CAS-Nummer: 1054483-36-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...