3-Bromo-6-(methoxymethoxy)-o-xylene

Lieferant: Apollo Scientific

OR470856-5G
APOSOR470856-5GEA 1120 EUR
APOSOR470856-5G
3-Bromo-6-(methoxymethoxy)-o-xylene
3-Bromo-6-(methoxymethoxy)-o-xylene
Formel: C₁₀H₁₃BrO₂
Molecular Weight: 245,113 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27935368
CAS-Nummer: 1301147-34-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR