Methyl 2-[(4-Amino-6-chloro-3-pyridyl)oxy]acetate

Lieferant: Apollo Scientific

OR470857-1G
APOSOR470857-1GEA 1330 EUR
APOSOR470857-1G
Methyl 2-[(4-Amino-6-chloro-3-pyridyl)oxy]acetate
Methyl 2-[(4-Amino-6-chloro-3-pyridyl)oxy]acetate
Formel: C₈H₉ClN₂O₃
Molecular Weight: 216,622 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27935367
CAS-Nummer: 1823872-37-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR