Methyl 2-[(6-Chloro-3-pyridyl)oxy]acetate

Lieferant: Apollo Scientific

OR470858-1G
APOSOR470858-1GEA 591 EUR
APOSOR470858-1G
Methyl 2-[(6-Chloro-3-pyridyl)oxy]acetate
Methyl 2-[(6-Chloro-3-pyridyl)oxy]acetate
Formel: C₈H₈ClNO₃
Molecular Weight: 201,607 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27933286
CAS-Nummer: 928118-49-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR