Methyl 3-(Methylsulphonamido)thiophene-2-carboxylate

Lieferant: Apollo Scientific

OR470859-5G
APOSOR470859-5GEA 713 EUR
APOSOR470859-5G
Methyl 3-(Methylsulphonamido)thiophene-2-carboxylate
Methyl 3-(Methylsulphonamido)thiophene-2-carboxylate
Formel: C₇H₉NO₄S₂
Molecular Weight: 235,281 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00126700
CAS-Nummer: 79128-71-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR