Methyl 3-[Bis(methylsulphonyl)amino]thiophene-2-carboxylate

Lieferant: Apollo Scientific

OR470860-5G
APOSOR470860-5GEA 713 EUR
APOSOR470860-5G
Methyl 3-[Bis(methylsulphonyl)amino]thiophene-2-carboxylate
Methyl 3-[Bis(methylsulphonyl)amino]thiophene-2-carboxylate
Formel: C₈H₁₁NO₆S₃
Molecular Weight: 313,371 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26381896
CAS-Nummer: 214532-29-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR