5-Amino-2-(aminomethyl)pyridine
Lieferant: Apollo Scientific
Synonyme:
6-(Aminomethyl)pyridin-3-amine
OR470861-1G
APOSOR470861-1GEA
916
EUR
APOSOR470861-1G
5-Amino-2-(aminomethyl)pyridine
5-Amino-2-(aminomethyl)pyridine
Formel:
C₆H₉N₃ Molecular Weight: 123,156 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06213791 CAS-Nummer: 771574-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...