tert-Butyl (2-methoxy-1,1-dimethylethyl)carbamate

Lieferant: Apollo Scientific

Synonyme: N-Boc-1-methoxy-2-methyl-2-propanamine

OR470862-5G
APOSOR470862-5GEA 804 EUR
APOSOR470862-5G
tert-Butyl (2-methoxy-1,1-dimethylethyl)carbamate
tert-Butyl (2-methoxy-1,1-dimethylethyl)carbamate
Formel: C₁₀H₂₁NO₃
Molecular Weight: 203,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15143199
CAS-Nummer: 204707-34-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR