5-Bromo-6-methyl-2-(methylthio)pyrimidin-4(3H)-one

Lieferant: Apollo Scientific

OR470863-1G
APOSOR470863-1GEA 570 EUR
APOSOR470863-1G
5-Bromo-6-methyl-2-(methylthio)pyrimidin-4(3H)-one
5-Bromo-6-methyl-2-(methylthio)pyrimidin-4(3H)-one
Formel: C₆H₇BrN₂OS
Molecular Weight: 235,102 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11617996
CAS-Nummer: 33238-63-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR