2-[2-(2-Methyl-1,3-dioxolan-2-yl)ethyl]isoindoline-1,3-dione

Lieferant: Apollo Scientific

OR470864-5G
APOSOR470864-5GEA 49.3 EUR
APOSOR470864-5G
2-[2-(2-Methyl-1,3-dioxolan-2-yl)ethyl]isoindoline-1,3-dione
2-[2-(2-Methyl-1,3-dioxolan-2-yl)ethyl]isoindoline-1,3-dione
Formel: C₁₄H₁₅NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09923388
CAS-Nummer: 84764-41-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR