Ethyl 1-Cyclopropyl-2,4-dioxopiperidine-3-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR470866-1G
APOSOR470866-1GEA
1010
EUR
APOSOR470866-1G
Ethyl 1-Cyclopropyl-2,4-dioxopiperidine-3-carboxylate
Ethyl 1-Cyclopropyl-2,4-dioxopiperidine-3-carboxylate
Formel:
C₁₁H₁₅NO₄ Molecular Weight: 225,241 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27935386 CAS-Nummer: 1453423-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...