4-Amino-2,6-dimethoxypyridine

Lieferant: Apollo Scientific

OR470868-1G
APOSOR470868-1GEA 1010 EUR
APOSOR470868-1G
4-Amino-2,6-dimethoxypyridine
4-Amino-2,6-dimethoxypyridine
Formel: C₇H₁₀N₂O₂
Molecular Weight: 154,166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19688654
CAS-Nummer: 17325-39-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR