2-(Boc-amino)-1-iodopropane

Lieferant: Apollo Scientific

OR470869-5G
APOSOR470869-5GEA 1010 EUR
APOSOR470869-5G
2-(Boc-amino)-1-iodopropane
2-(Boc-amino)-1-iodopropane
Formel: C₈H₁₆INO₂
Molecular Weight: 285.123 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17214622
CAS-Nummer: 363599-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR