Methyl 3-(4-Ethynylphenyl)propanoate

Lieferant: Apollo Scientific
Achtung

OR470870-1G
APOSOR470870-1GEA 819 EUR
APOSOR470870-1G
Methyl 3-(4-Ethynylphenyl)propanoate
Methyl 3-(4-Ethynylphenyl)propanoate
Formel: C₁₂H₁₂O₂
Molecular Weight: 188,222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26398532
CAS-Nummer: 1068471-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR