6-Chloro-4-hydroxy-2-(methylthio)pyrimidine

Lieferant: Apollo Scientific
Gefahr

OR470872-5G
APOSOR470872-5GEA 713 EUR
APOSOR470872-5G
6-Chloro-4-hydroxy-2-(methylthio)pyrimidine
6-Chloro-4-hydroxy-2-(methylthio)pyrimidine
Formel: C₅H₅ClN₂OS
Molecular Weight: 176,624 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16875711
CAS-Nummer: 6632-63-9
UN: 3263
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR