tert-Butyl-2-(hydroxymethyl)-1-oxa-7-azaspiro[3.5]nonane-7-carboxylate

Lieferant: Apollo Scientific

Synonyme: 7-Boc-2-(hydroxymethyl)-1-oxa-7-azaspiro[3.5]nonane

OR470873-1G
APOSOR470873-1GEA 1500 EUR
APOSOR470873-1G
tert-Butyl-2-(hydroxymethyl)-1-oxa-7-azaspiro[3.5]nonane-7-carboxylate
tert-Butyl-2-(hydroxymethyl)-1-oxa-7-azaspiro[3.5]nonane-7-carboxylate
Formel: C₁₃H₂₃NO₄
Molecular Weight: 257,33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13184495
CAS-Nummer: 203662-53-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR