1-Isopropylpiperidine-2,4-dione
Lieferant: Apollo Scientific
OR470874-1G
APOSOR470874-1GEA
916
EUR
APOSOR470874-1G
1-Isopropylpiperidine-2,4-dione
1-Isopropylpiperidine-2,4-dione
Formel:
C₈H₁₃NO₂ Molecular Weight: 155,194 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21944001 CAS-Nummer: 904301-91-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...