2-(Boc-amino)-5-chloro-3-hydroxypyridine

Lieferant: Apollo Scientific

OR470877-1G
APOSOR470877-1GEA 0 EUR
APOSOR470877-1G
2-(Boc-amino)-5-chloro-3-hydroxypyridine
2-(Boc-amino)-5-chloro-3-hydroxypyridine
Formel: C₁₀H₁₃ClN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD27935389
CAS-Nummer: 1609402-46-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR