2-(Boc-amino)-5-bromo-3-hydroxypyridine

Lieferant: Apollo Scientific

Synonyme: tert-Butyl (5-bromo-3-hydroxypyridin-2-yl)carbamate

OR470878-1G
APOSOR470878-1GEA 91.5 EUR
APOSOR470878-1G
2-(Boc-amino)-5-bromo-3-hydroxypyridine
2-(Boc-amino)-5-bromo-3-hydroxypyridine
Formel: C₁₀H₁₃BrN₂O₃
Molecular Weight: 289.126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12756114
CAS-Nummer: 1207175-73-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR