2-Hydroxy-7-methoxy-1-naphthaldehyde

Lieferant: Apollo Scientific

OR470880-1G
APOSOR470880-1GEA 916 EUR
APOSOR470880-1G
2-Hydroxy-7-methoxy-1-naphthaldehyde
2-Hydroxy-7-methoxy-1-naphthaldehyde
Formel: C₁₂H₁₀O₃
Molecular Weight: 202,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22416262
CAS-Nummer: 75965-66-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR