cis-3,7-Diazabicyclo[3.3.0]octane
Lieferant: Apollo Scientific
OR470881-1G
APOSOR470881-1GEA
1010
EUR
APOSOR470881-1G
cis-3,7-Diazabicyclo[3.3.0]octane
cis-3,7-Diazabicyclo[3.3.0]octane
Formel:
C₆H₁₂N₂ Molecular Weight: 112,173 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27935365 CAS-Nummer: 19885-60-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...