1-(2-Acetoxyethyl)pyrazole-4-boronic acid pinacol ester

Lieferant: Apollo Scientific

OR470884-1G
APOSOR470884-1GEA 1010 EUR
APOSOR470884-1G
1-(2-Acetoxyethyl)pyrazole-4-boronic acid pinacol ester
1-(2-Acetoxyethyl)pyrazole-4-boronic acid pinacol ester
Formel: C₁₃H₂₁BN₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD14707223
CAS-Nummer: 1251731-71-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR