5-(Aminomethyl)-2-chlorophenylboronic acid pinacol ester

Lieferant: Apollo Scientific

OR470886-1G
APOSOR470886-1GEA 1540 EUR
APOSOR470886-1G
5-(Aminomethyl)-2-chlorophenylboronic acid pinacol ester
5-(Aminomethyl)-2-chlorophenylboronic acid pinacol ester
Formel: C₁₃H₁₉BClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18730170
CAS-Nummer: 1544673-71-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR