5-(Aminomethyl)-2-methylphenylboronic acid pinacol ester

Lieferant: Apollo Scientific

OR470887-1G
APOSOR470887-1GEA 1540 EUR
APOSOR470887-1G
5-(Aminomethyl)-2-methylphenylboronic acid pinacol ester
5-(Aminomethyl)-2-methylphenylboronic acid pinacol ester
Formel: C₁₄H₂₂BNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18730174
CAS-Nummer: 1011460-69-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR