3,6-Dichloro-4-iodopyridazine

Lieferant: Apollo Scientific

OR470888-5G
APOSOR470888-5GEA 1300 EUR
APOSOR470888-5G
3,6-Dichloro-4-iodopyridazine
3,6-Dichloro-4-iodopyridazine
Formel: C₄HCl₂IN₂
Molecular Weight: 274,875 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18249972
CAS-Nummer: 130825-13-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR