N,N-Diethyl-1,3-benzenediamine hydrochloride

Lieferant: Apollo Scientific

OR470891-5G
APOSOR470891-5GEA 1010 EUR
APOSOR470891-5G
N,N-Diethyl-1,3-benzenediamine hydrochloride
N,N-Diethyl-1,3-benzenediamine hydrochloride
Formel: C₁₀H₁₆N₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD27959538
CAS-Nummer: 214707-03-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR