2,2-Dimethyl-2H-chromene-6-carbaldehyde

Lieferant: Apollo Scientific

OR470894-1G
APOSOR470894-1GEA 804 EUR
APOSOR470894-1G
2,2-Dimethyl-2H-chromene-6-carbaldehyde
2,2-Dimethyl-2H-chromene-6-carbaldehyde
Formel: C₁₂H₁₂O₂
Molecular Weight: 188,222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17215738
CAS-Nummer: 69964-40-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR