Methyl 2-(6-Hydroxybenzofuran-3-yl)acetate 97%

Lieferant: Apollo Scientific
Achtung

OR470895-1G
APOSOR470895-1GEA 295 EUR
APOSOR470895-1G
Methyl 2-(6-Hydroxybenzofuran-3-yl)acetate 97%
Methyl 2-(6-Hydroxybenzofuran-3-yl)acetate
Formel: C₁₁H₁₀O₄
Molecular Weight: 206,195 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16658283
CAS-Nummer: 726174-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR