3-Bromo-4-methoxybenzylamine

Lieferant: Apollo Scientific

OR470896-5G
APOSOR470896-5GEA 1010 EUR
APOSOR470896-5G
3-Bromo-4-methoxybenzylamine
3-Bromo-4-methoxybenzylamine
Formel: C₈H₁₀BrNO
Molecular Weight: 216.075 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03410965
CAS-Nummer: 247254-47-3
UN: 2735
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR