3-Bromo-4-methylbenzylamine
Lieferant: Apollo Scientific
OR470897-1G
APOSOR470897-1GEA
954
EUR
APOSOR470897-1G
3-Bromo-4-methylbenzylamine
3-Bromo-4-methylbenzylamine
Formel:
C₈H₁₀BrN Molecular Weight: 200,076 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18392011 CAS-Nummer: 1177558-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...