3-(4-Chlorobenzyl)-2-piperidone

Lieferant: Apollo Scientific

OR470898-1G
APOSOR470898-1GEA 916 EUR
APOSOR470898-1G
3-(4-Chlorobenzyl)-2-piperidone
3-(4-Chlorobenzyl)-2-piperidone
Formel: C₁₂H₁₄ClNO
Molecular Weight: 223,699 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11848670
CAS-Nummer: 1260763-19-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR