3-(2-Chlorobenzyl)-2-piperidone

Lieferant: Apollo Scientific

OR470899-1G
APOSOR470899-1GEA 916 EUR
APOSOR470899-1G
3-(2-Chlorobenzyl)-2-piperidone
3-(2-Chlorobenzyl)-2-piperidone
Formel: C₁₂H₁₄ClNO
Molecular Weight: 223,699 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11848668
CAS-Nummer: 1260766-37-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR