2-[4-(2-Aminoethyl)phenoxy]acetic acid
Lieferant: Apollo Scientific
OR470900-5G
APOSOR470900-5GEA
1330
EUR
APOSOR470900-5G
2-[4-(2-Aminoethyl)phenoxy]acetic acid
2-[4-(2-Aminoethyl)phenoxy]acetic acid
Formel:
C₁₀H₁₃NO₃ Molecular Weight: 195,215 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08448865 CAS-Nummer: 55458-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...