2-(4-Cyclobutoxyphenyl)ethylamine

Lieferant: Apollo Scientific

OR470902-1G
APOSOR470902-1GEA 916 EUR
APOSOR470902-1G
2-(4-Cyclobutoxyphenyl)ethylamine
2-(4-Cyclobutoxyphenyl)ethylamine
Formel: C₁₂H₁₇NO
Lagertemperatur: Raumtemperatur
MDL: MFCD23107401
CAS-Nummer: 1554206-32-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR