2-[4-(2-Cyclopropylethoxy)phenyl]ethylamine

Lieferant: Apollo Scientific

OR470903-1G
APOSOR470903-1GEA 1120 EUR
APOSOR470903-1G
2-[4-(2-Cyclopropylethoxy)phenyl]ethylamine
2-[4-(2-Cyclopropylethoxy)phenyl]ethylamine
Formel: C₁₃H₁₉NO
Molecular Weight: 205,296 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21260218
CAS-Nummer: 1481230-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR