2-[4-(Cyclohexyloxy)phenyl]ethylamine
Lieferant: Apollo Scientific
OR470904-1G
APOSOR470904-1GEA
916
EUR
APOSOR470904-1G
2-[4-(Cyclohexyloxy)phenyl]ethylamine
2-[4-(Cyclohexyloxy)phenyl]ethylamine
Formel:
C₁₄H₂₁NO Molecular Weight: 219,323 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11167592 CAS-Nummer: 1042579-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...