2-[4-(Tetrahydropyran-2-yloxy)phenyl]ethylamine

Lieferant: Apollo Scientific

OR470905-1G
APOSOR470905-1GEA 713 EUR
APOSOR470905-1G
2-[4-(Tetrahydropyran-2-yloxy)phenyl]ethylamine
2-[4-(Tetrahydropyran-2-yloxy)phenyl]ethylamine
Formel: C₁₃H₁₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09924455
CAS-Nummer: 1000550-74-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR