2-[4-(Pyrrolidin-3-yloxy)phenyl]ethylamine

Lieferant: Apollo Scientific

OR470906-1G
APOSOR470906-1GEA 1220 EUR
APOSOR470906-1G
2-[4-(Pyrrolidin-3-yloxy)phenyl]ethylamine
2-[4-(Pyrrolidin-3-yloxy)phenyl]ethylamine
Formel: C₁₂H₁₈N₂O
Molecular Weight: 206,284 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09924818
CAS-Nummer: 1000507-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR