2-[4-(Pyridin-2-ylmethoxy)phenyl]ethylamine

Lieferant: Apollo Scientific

OR470907-1G
APOSOR470907-1GEA 916 EUR
APOSOR470907-1G
2-[4-(Pyridin-2-ylmethoxy)phenyl]ethylamine
2-[4-(Pyridin-2-ylmethoxy)phenyl]ethylamine
Formel: C₁₄H₁₆N₂O
Molecular Weight: 228,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09050953
CAS-Nummer: 663597-30-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR