2-[4-(Pyridin-3-ylmethoxy)phenyl]ethylamine

Lieferant: Apollo Scientific

OR470908-1G
APOSOR470908-1GEA 713 EUR
APOSOR470908-1G
2-[4-(Pyridin-3-ylmethoxy)phenyl]ethylamine
2-[4-(Pyridin-3-ylmethoxy)phenyl]ethylamine
Formel: C₁₄H₁₆N₂O
Molecular Weight: 228,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09044240
CAS-Nummer: 1343316-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR