1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyne

Lieferant: Apollo Scientific

Synonyme: Ethyne-1,2-diyl bis(boronic acid Pinacol ester), Acetylene-1,2-diyl bis(boronic acid Pinacol ester)

OR470909-1G
APOSOR470909-1GEA 1290 EUR
APOSOR470909-1G
1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyne
1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyne
Formel: C₁₄H₂₄B₂O₄
Molecular Weight: 277,96 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22666429
CAS-Nummer: 1010840-17-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR