Methyl 2-(7-Hydroxybenzofuran-3-yl)acetate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-(7-hydroxybenzofuran-3-yl)acetate

OR470910-1G
APOSOR470910-1GEA 1190 EUR
APOSOR470910-1G
Methyl 2-(7-Hydroxybenzofuran-3-yl)acetate
Methyl 2-(7-Hydroxybenzofuran-3-yl)acetate
Formel: C₁₁H₁₀O₄
Molecular Weight: 206,195 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09991503
CAS-Nummer: 181052-63-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR