2-[3-(Methylsulphonyl)phenyl]ethylamine hydrochloride

Lieferant: Apollo Scientific

OR470915-5G
APOSOR470915-5GEA 1010 EUR
APOSOR470915-5G
2-[3-(Methylsulphonyl)phenyl]ethylamine hydrochloride
2-[3-(Methylsulphonyl)phenyl]ethylamine hydrochloride
Formel: C₉H₁₃NO₂S·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD27986815
CAS-Nummer: 1073666-57-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR