2-[2-(Methylsulphonyl)phenyl]acetic acid

Lieferant: Apollo Scientific

OR470917-5G
APOSOR470917-5GEA 1330 EUR
APOSOR470917-5G
2-[2-(Methylsulphonyl)phenyl]acetic acid
2-[2-(Methylsulphonyl)phenyl]acetic acid
Formel: C₉H₁₀O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD11898924
CAS-Nummer: 142336-20-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR