4-(Dimethylcarbamoyl)-2-methylphenylboronic acid

Lieferant: Apollo Scientific

Synonyme: 4-(Dimethylcarbamoyl)-2-methylphenylboronic Acid

OR470918-1G
APOSOR470918-1GEA 1010 EUR
APOSOR470918-1G
4-(Dimethylcarbamoyl)-2-methylphenylboronic acid
4-(Dimethylcarbamoyl)-2-methylphenylboronic acid
Formel: C₁₀H₁₄BNO₃
Molecular Weight: 207,034 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27977108
CAS-Nummer: 159306-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR