N-Boc-2-(1-Iminoethyl)hydrazine

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 2-(1-iminoethyl)hydrazine-1-carboxylate

OR470919-1G
APOSOR470919-1GEA 253 EUR
APOSOR470919-1G
N-Boc-2-(1-Iminoethyl)hydrazine
N-Boc-2-(1-Iminoethyl)hydrazine
Formel: C₇H₁₅N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD27986814
CAS-Nummer: 851535-08-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR