5-Aminothiazolo[4,5-d]pyrimidin-2(3H)-one

Lieferant: Apollo Scientific

OR470920-1G
APOSOR470920-1GEA 484 EUR
APOSOR470920-1G
5-Aminothiazolo[4,5-d]pyrimidin-2(3H)-one
5-Aminothiazolo[4,5-d]pyrimidin-2(3H)-one
Formel: C₅H₄N₄OS
Molecular Weight: 168,176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17017931
CAS-Nummer: 848691-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR